Please login to your online account to display your discounted pricing

4-Hydrazinobenzene-1-sulfonamide hydrochloride, Maybridge


Quantity 100g

Manufacturer: fisher scientific company  BTB06645FA

 View more versions of this product

Catalog No. BTB06645FA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 223.68
Quantity 100g
Assay 95%
CAS Max % 100.0
Linear Formula NH2NHC6H4SO2NH2·HCl
Molecular Formula C6H9N3O2S·ClH
CAS Min % 94.5
MDL Number MFCD00052262
Assay Percent Range 95%
Chemical Name or Material 4-Hydrazinobenzene-1-sulfonamide hydrochloride
CAS 17852-52-7

Specifications

Chemical Name or Material 4-Hydrazinobenzene-1-sulfonamide hydrochloride
CAS 17852-52-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H9N3O2S·ClH
Linear Formula NH2NHC6H4SO2NH2·HCl
Formula Weight 223.68
MDL Number MFCD00052262