Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hexyn-3-ol, 95%

Chemical Identifiers

Chemical Name or Material 4-Hexyn-3-ol
CAS 20739-59-7
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00041591
UN Number UN1987
Beilstein 1739392
Synonym Ethyl 1-propynyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0972706

  SDS
alfa aesar™
L0972706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0972714

  SDS
alfa aesar™
L0972714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
MDL Number MFCD00041591
CAS 20739-59-7
Molecular Formula C6H10O
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 1 Warning
UN Number UN1987
Synonym Ethyl 1-propynyl carbinol
Assay 95%
Beilstein 1739392
Quantity 5g
Boiling Point 94° to 95°C (105mmHg)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Chemical Name or Material 4-Hexyn-3-ol
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.