missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hexyn-3-ol, 95%

Manufacturer:  Alfa Aesar™ L0972714

 View more versions of this product

Catalog No. AAL0972714



Specifications

Specifications

4-Hexyn-3-ol
25g
C6H10O
UN1987
4-hexyn-3-ol, ethyl 1-propynyl carbinol, 1-pentyn-3-ol, methyl, 4-hexyne-3-ol, acmc-1cpld, 4-hexyn-3-ol, 4-hexyn-3-ol
CCC(C#CC)O
98.145
98.15
94°C to 95°C (105mmHg)
20739-59-7
MFCD00041591
1739392
KARLLBDFLHNKBO-UHFFFAOYSA-N
hex-4-yn-3-ol
140751
95%

Chemical Identifiers

20739-59-7
98.145
KARLLBDFLHNKBO-UHFFFAOYSA-N
140751
CCC(C#CC)O
C6H10O
MFCD00041591
4-hexyn-3-ol, ethyl 1-propynyl carbinol, 1-pentyn-3-ol, methyl, 4-hexyne-3-ol, acmc-1cpld, 4-hexyn-3-ol, 4-hexyn-3-ol
hex-4-yn-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

Documents