missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hexylphenol, TCI America™

$135.97 - $409.45

Chemical Identifiers

CAS 2446-69-7
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00051313
InChI Key SZWBRVPZWJYIHI-UHFFFAOYSA-N
Synonym 4-n-hexylphenol, phenol, 4-hexyl, p-hexylphenol, p-n-hexylphenol, phenol,4-hexyl, unii-3v1ke0n8d5, phenol, p-hexyl, acmc-209gch, 1-4-hydroxyphenyl hexane, bidd:er0077
PubChem CID 17132
ChEBI CHEBI:34439
IUPAC Name 4-hexylphenol
SMILES CCCCCCC1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H06115G View Documents TCI America
H06115G
5g Each for $135.97
 
H061125G View Documents TCI America
H061125G
25g Each for $409.45
 
Specifications

Chemical Identifiers

2446-69-7
178.275
SZWBRVPZWJYIHI-UHFFFAOYSA-N
17132
4-hexylphenol
C12H18O
MFCD00051313
4-n-hexylphenol, phenol, 4-hexyl, p-hexylphenol, p-n-hexylphenol, phenol,4-hexyl, unii-3v1ke0n8d5, phenol, p-hexyl, acmc-209gch, 1-4-hydroxyphenyl hexane, bidd:er0077
CHEBI:34439
CCCCCCC1=CC=C(C=C1)O

Specifications

White-Yellow
2446-69-7
MFCD00051313
4-n-hexylphenol, phenol, 4-hexyl, p-hexylphenol, p-n-hexylphenol, phenol,4-hexyl, unii-3v1ke0n8d5, phenol, p-hexyl, acmc-209gch, 1-4-hydroxyphenyl hexane, bidd:er0077
CCCCCCC1=CC=C(C=C1)O
178.275
CHEBI:34439
Crystal-Lump at 20°C
4-Hexylphenol
5g
C12H18O
2430
SZWBRVPZWJYIHI-UHFFFAOYSA-N
4-hexylphenol
17132
178.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes