Please login to your online account to display your discounted pricing

4-Hexyloxybenzaldehyde 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  411671000

 View more versions of this product

Catalog No. AC411671000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 206.28
Quantity 100g
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 227-251-0
Density 0.992g/mL
CAS Max % 100.0
Refractive Index 1.5285 to 1.5325 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 99%
Molecular Formula C13H18O2
CAS 5736-94-7
Assay Percent Range 99%
Chemical Name or Material 4-Hexyloxybenzaldehyde
MDL Number MFCD00016615

Specifications

Chemical Name or Material 4-Hexyloxybenzaldehyde
Name Note 99%
CAS 5736-94-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H18O2
Formula Weight 206.28
MDL Number MFCD00016615
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents