missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hexyloxybenzaldehyde 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 411670250

 View more versions of this product

Catalog No. AC411670250


 

Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Clear colorless to light yellow
100.0
98.5
Authentic
C13H18O2
MFCD00016615
99%
206.28
25g
0.992g/mL
1.5285 to 1.5325 (20°C, 589nm)
Glass bottle
4-Hexyloxybenzaldehyde
5736-94-7
99%

Chemical Identifiers

5736-94-7
100.0
C13H18O2
MFCD00016615
≥98.5%
98.5
99%
206.28
4-Hexyloxybenzaldehyde, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-251-0

TSCA : TSCA