Please login to your online account to display your discounted pricing

4-Hexen-3-one 98%, mixture of isomers, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Hexen-3-one
CAS 2497-21-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (sum of isomers)
Assay Percent Range 98%
Molecular Formula C6H10O
Linear Formula CH3CH=CHC(O)CH2CH3
Formula Weight 98.14
MDL Number MFCD00010631
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391700050

  SDS
thermo fisher scientific
AC391700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (sum of isomers)
Physical Form Liquid
Formula Weight 98.14
Quantity 5g
Color Clear colorless to yellow
Density 0.8580g/mL
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H10O
Linear Formula CH3CH=CHC(O)CH2CH3
MDL Number MFCD00010631
CAS 2497-21-4
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Hexen-3-one
Infrared Spectrum Authentic
Refractive Index 1.4350 to 1.4450 (20°C, 589nm)
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.