missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hexen-1-ol, predominantly trans, 97%

Manufacturer:  Alfa Aesar™ H5445906

 View more versions of this product

Catalog No. AAH5445906



Description

Description

Applications
4-Hexen-1-ol on reaction with NaIO4/NaHSO3 reagent yields 2-(1-iodoethyl)tetrahydrofuran. has been employed in the preparation of hex-4-enals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

4-Hexen-1-ol
61°C (142°F)
159°C to 160°C
predominantly trans
C6H12O
MFCD00009713
Slightly soluble in water.
CC=CCCCO
100.161
100.16
0.851
1.439
5g
928-92-7
CH3CH=CH(CH2)3OH
4-hexen-1-ol
VTIODUHBZHNXFP-NSCUHMNNSA-N
(E)-hex-4-en-1-ol
641248
97%

Chemical Identifiers

928-92-7
100.161
VTIODUHBZHNXFP-NSCUHMNNSA-N
641248
CC=CCCCO
C6H12O
MFCD00009713
4-hexen-1-ol
(E)-hex-4-en-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P264b-P280-P305+P351+P338-P370+P378q-P501c

H227-H319

EINECSNumber : 213-188-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents