missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hexen-1-ol, 97%, predominantly trans, Thermo Scientific™
Chemical Identifiers
CAS | 928-92-7 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.16 |
MDL Number | MFCD00009713 |
InChI Key | VTIODUHBZHNXFP-NSCUHMNNSA-N |
Synonym | 4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans |
PubChem CID | 641248 |
IUPAC Name | (E)-hex-4-en-1-ol |
SMILES | C\C=C\CCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC217950050
|
Thermo Scientific Chemicals
217950050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
928-92-7 | |
100.16 | |
VTIODUHBZHNXFP-NSCUHMNNSA-N | |
641248 | |
C\C=C\CCCO |
C6H12O | |
MFCD00009713 | |
4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans | |
(E)-hex-4-en-1-ol |
Specifications
928-92-7 | |
100.0 | |
159.0°C to 160.0°C | |
Authentic | |
Glass bottle | |
1.4394 | |
MFCD00009713 | |
01,III,1929 | |
4-hexen-1-ol, e-4-hexen-1-ol, e-hex-4-en-1-ol, trans-4-hexen-1-ol, 4-hexen-1-ol, 4e, 4-hexen-1-ol, e, trans-4-hexenol, unii-zhb2532c2h, 4-hexen-1-ol, predominantly trans | |
C\C=C\CCCO | |
100.16 | |
100.16 | |
4-Hexen-1-ol |
96.0 | |
0.8500g/mL | |
61°C | |
96% min. (GC) | |
C6H12O | |
CH3CH=CH(CH2)3OH | |
5g | |
0.85 | |
VTIODUHBZHNXFP-NSCUHMNNSA-N | |
(E)-hex-4-en-1-ol | |
641248 | |
97% |
Safety and Handling
GHS H Statement:
GHS P Statement
Wear eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 213-188-6
RUO â Research Use Only