Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Hexamethyleneiminomethyl)benzeneboronic acid pinacol ester, 95%

$327.78 - $327.78

Chemical Identifiers

Chemical Name or Material 4-(Hexamethyleneiminomethyl)benzeneboronic acid pinacol ester
CAS 1315278-37-5
Molecular Formula C19H30BNO2
Formula Weight 315.26g/mol
MDL Number MFCD16294503
Synonym 4-(1-Azepanylmethyl)benzeneboronic acid pinacol ester; 4-(1-Homopiperidinylmethyl)benzeneboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5174406

 
alfa aesar™
H51744-06
Each for $327.78
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 4-(Hexamethyleneiminomethyl)benzeneboronic acid pinacol ester
CAS 1315278-37-5
Assay 95%
Molecular Formula C19H30BNO2
Formula Weight 315.26g/mol
MDL Number MFCD16294503
Synonym 4-(1-Azepanylmethyl)benzeneboronic acid pinacol ester; 4-(1-Homopiperidinylmethyl)benzeneboronic acid pinacol ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.