missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Heptyn-2-ol, 97%

Manufacturer:  Alfa Aesar™ L1129214

 View more versions of this product

Catalog No. AAL1129214



Specifications

Specifications

4-Heptyn-2-ol
67°C to 68°C (14mmHg)
19781-81-8
MFCD00041588
4-heptyn-2-ol, acmc-1c9g1, cxviebmewksony-uhfffaoysa
CCC#CCC(C)O
112.172
112.17
62°C (143°F)
25g
C7H12O
2232633
CXVIEBMEWKSONY-UHFFFAOYSA-N
hept-4-yn-2-ol
89225
97%

Chemical Identifiers

19781-81-8
112.172
CXVIEBMEWKSONY-UHFFFAOYSA-N
89225
CCC#CCC(C)O
C7H12O
MFCD00041588
4-heptyn-2-ol, acmc-1c9g1, cxviebmewksony-uhfffaoysa
hept-4-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents