missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Heptylbenzoyl Chloride, TCI America™

$177.22 - $605.02

Chemical Identifiers

CAS 50606-96-7
Molecular Formula C14H19ClO
Molecular Weight (g/mol) 238.755
MDL Number MFCD00000702
InChI Key WHTFLTOKFXTJGV-UHFFFAOYSA-N
Synonym 4-n-heptylbenzoyl chloride, benzoyl chloride, 4-heptyl, p-heptylbenzoyl chloride, acmc-209ko7, benzoyl chloride,4-heptyl, 4-n-heptyl-benzoyl chloride, 4-heptylbenzoyl chloride, 4-n-heptyl benzoic acid chloride, 4-hept-1-yl benzoyl chloride 95+%
PubChem CID 170864
IUPAC Name 4-heptylbenzoyl chloride
SMILES CCCCCCCC1=CC=C(C=C1)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07925G View Documents TCI America
H07925G
5g Each for $177.22
 
H079225G View Documents TCI America
H079225G
25g Each for $605.02
 
Specifications

Chemical Identifiers

50606-96-7
238.755
WHTFLTOKFXTJGV-UHFFFAOYSA-N
170864
CCCCCCCC1=CC=C(C=C1)C(=O)Cl
C14H19ClO
MFCD00000702
4-n-heptylbenzoyl chloride, benzoyl chloride, 4-heptyl, p-heptylbenzoyl chloride, acmc-209ko7, benzoyl chloride,4-heptyl, 4-n-heptyl-benzoyl chloride, 4-heptylbenzoyl chloride, 4-n-heptyl benzoic acid chloride, 4-hept-1-yl benzoyl chloride 95+%
4-heptylbenzoyl chloride

Specifications

196°C
5g
C14H19ClO
3265
WHTFLTOKFXTJGV-UHFFFAOYSA-N
4-heptylbenzoyl chloride
170864
Clear Liquid at 20°C
4-Heptylbenzoyl Chloride
Yellow
50606-96-7
MFCD00000702
4-n-heptylbenzoyl chloride, benzoyl chloride, 4-heptyl, p-heptylbenzoyl chloride, acmc-209ko7, benzoyl chloride,4-heptyl, 4-n-heptyl-benzoyl chloride, 4-heptylbenzoyl chloride, 4-n-heptyl benzoic acid chloride, 4-hept-1-yl benzoyl chloride 95+%
CCCCCCCC1=CC=C(C=C1)C(=O)Cl
238.755
238.76
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes