missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Heptylbenzonitrile, TCI America™

$94.72 - $241.39

Chemical Identifiers

CAS 60484-67-5
Molecular Formula C14H19N
Molecular Weight (g/mol) 201.313
MDL Number MFCD00143348
InChI Key XTIKBCXMOYZUMG-UHFFFAOYSA-N
Synonym 1-(4-Cyanophenyl)heptane
PubChem CID 575863
IUPAC Name 4-heptylbenzonitrile
SMILES CCCCCCCC1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07785ML View Documents TCI America
H07785ML
5mL Each for $241.39
 
H07781ML View Documents TCI America
H07781ML
1mL Each for $94.72
 
Specifications

Chemical Identifiers

60484-67-5
201.313
XTIKBCXMOYZUMG-UHFFFAOYSA-N
575863
CCCCCCCC1=CC=C(C=C1)C#N
C14H19N
MFCD00143348
1-(4-Cyanophenyl)heptane
4-heptylbenzonitrile

Specifications

Yellow
60484-67-5
MFCD00143348
XTIKBCXMOYZUMG-UHFFFAOYSA-N
4-heptylbenzonitrile
575863
Clear Liquid at 20°C
4-Heptylbenzonitrile
5mL
C14H19N
1-(4-Cyanophenyl)heptane
CCCCCCCC1=CC=C(C=C1)C#N
201.313
201.31
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3541

TSCA : No