missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Guanidinobenzoic acid hydrochloride, 99%

$41.78 - $171.18

Chemical Identifiers

CAS 42823-46-1
Molecular Formula C8H10ClN3O2
Molecular Weight (g/mol) 215.637
MDL Number MFCD00040587
InChI Key YETFLAUJROGBMC-UHFFFAOYSA-N
Synonym 4-guanidinobenzoic acid hydrochloride, n-4-carboxyphenyl guanidine hydrochloride, 4-guanidinobenzoic acid hcl, 4-guanidino benzoic acid hydrochloride, 4-amidinoamino benzoic acid, chloride, pubchem14981, pubchem14993, acmc-1an3m, ksc494a0j, 4-guanidino-benzoic acid-hcl salt
PubChem CID 3084875
IUPAC Name 4-(diaminomethylideneamino)benzoic acid;hydrochloride
SMILES C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6039203 View Documents Alfa Aesar™
H6039203
1g Each for $41.78
 
AAH6039206 View Documents Alfa Aesar™
H6039206
5g Each for $171.18
 
Specifications

Chemical Identifiers

42823-46-1
215.637
YETFLAUJROGBMC-UHFFFAOYSA-N
3084875
C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl
C8H10ClN3O2
MFCD00040587
4-guanidinobenzoic acid hydrochloride, n-4-carboxyphenyl guanidine hydrochloride, 4-guanidinobenzoic acid hcl, 4-guanidino benzoic acid hydrochloride, 4-amidinoamino benzoic acid, chloride, pubchem14981, pubchem14993, acmc-1an3m, ksc494a0j, 4-guanidino-benzoic acid-hcl salt
4-(diaminomethylideneamino)benzoic acid;hydrochloride

Specifications

285°C
42823-46-1
H2NC(=NH)NHC6H4CO2H·HCl
3718032
YETFLAUJROGBMC-UHFFFAOYSA-N
4-(diaminomethylideneamino)benzoic acid;hydrochloride
3084875
99%
1g
C8H10ClN3O2
MFCD00040587
4-guanidinobenzoic acid hydrochloride, n-4-carboxyphenyl guanidine hydrochloride, 4-guanidinobenzoic acid hcl, 4-guanidino benzoic acid hydrochloride, 4-amidinoamino benzoic acid, chloride, pubchem14981, pubchem14993, acmc-1an3m, ksc494a0j, 4-guanidino-benzoic acid-hcl salt
C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl
215.637
215.64
4-Guanidinobenzoic acid hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C