missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Guanidinobenzoic acid hydrochloride, 95%, ACROS Organics™

Chemical Identifiers

CAS 42823-46-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8 H9 N3 O2 . H Cl
Linear Formula NH2C(NH)NHC6H4COOH
Formula Weight 215.64
MDL Number MFCD00040587
Chemical Name or Material 4-Guanidinobenzoic acid hydrochloride, 95%
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC338310010 SDS Thermo Fisher Scientific
338310010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

42823-46-1
100.0
C8 H9 N3 O2 . H Cl
215.64
4-Guanidinobenzoic acid hydrochloride, 95%
94.0
95%
NH2C(NH)NHC6H4COOH
MFCD00040587
≥94%

Specifications

42823-46-1
100.0
Powder
Glass bottle
NH2C(NH)NHC6H4COOH
MFCD00040587
1g
≥94%
94.0
Authentic
95%
C8 H9 N3 O2 . H Cl
215.64
White to light beige
4-Guanidinobenzoic acid hydrochloride, 95%

Safety and Handling

EINECSNumber : 255-956-3