Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Furfuryliminomethyl)benzeneboronic acid pinacol ester, 97%

$475.35 - $475.35

Chemical Identifiers

Chemical Name or Material 4-(Furfuryliminomethyl)benzeneboronic acid pinacol ester
CAS 1218790-46-5
Molecular Formula C18H22BNO3
Formula Weight 311.18g/mol
MDL Number MFCD11113038
Synonym 1-[(2-Furylmethyl)iminomethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5112114

 
alfa aesar™
H51121-14
25g Each for $475.35
Description & Specifications

Specifications

Quantity 25g
Quantity 25g
Chemical Name or Material 4-(Furfuryliminomethyl)benzeneboronic acid pinacol ester
CAS 1218790-46-5
Assay 97%
Molecular Formula C18H22BNO3
Formula Weight 311.18g/mol
MDL Number MFCD11113038
Synonym 1-[(2-Furylmethyl)iminomethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene