Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

$64.49 - $198.89

Chemical Identifiers

Chemical Name or Material 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1073353-59-9
Molecular Formula C18H22BNO4
Formula Weight 327.18
MDL Number MFCD09266168
Synonym 4-(Furfurylaminocarbonyl)phenylboronic acid pinacol ester; N-Furfuryl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2923603

  SDS
alfa aesar™
H2923603
1g Each for $64.49

AAH2923606

  SDS
alfa aesar™
H2923606
5g Each for $198.89
Description & Specifications

Specifications

Formula Weight 327.18
Molecular Formula C18H22BNO4
Quantity 1g
Melting Point 113° to 114°C
Assay 97%
Synonym 4-(Furfurylaminocarbonyl)phenylboronic acid pinacol ester; N-Furfuryl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
MDL Number MFCD09266168
Chemical Name or Material 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1073353-59-9