missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 4 ft. Cell Logic™ Class II, Type B2 Biosafety Cabinet, US Model

Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.

$48137.02 - $48137.02

Specifications

Description 4 ft. Cell Logic Class II, Type B2 Biosafety Cabinet
Certifications/Compliance ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel (304)
Width (Metric) Interior 123.2 cm
Product Type Class II, Type B2 Bio Safety Cabinet
View More Specs

Products 1
Catalog Number Mfr. No. Includes Plug Type Price Quantity  
Catalog Number Mfr. No. Includes Plug Type Price Quantity  
01-812-449
View Documents
Labconco™
343481102
Cabinet, Accessory Package, Scope-Ready, Temp-Zone, Base Stand North America (115 V, 15 A) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Performance Features:

  • Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection.
    • Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow – even as HEPA filters load.
    • Optimized airflow minimizes energy use and noise output to industry-leading levels.
  • Logic OS with 5 in. line-of-sight touchscreen display.
    • Access cabinet information such as filter life, alerts and alarms, data logs.
    • Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
  • Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output.
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals.
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
  • Full, six-year warranty on all Class II biosafety cabinets.

Optional Configurations:

  • Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
    • Stand-Still™ Isolation Platform is required and sold separately.
  • Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
    • Water circulator is required (not included).
  • Telescoping base stand with shelf and leveling feet.
Specifications

Specifications

4 ft. Cell Logic Class II, Type B2 Biosafety Cabinet
Stainless Steel (304)
Class II, Type B2 Bio Safety Cabinet
61 dBA
105 W
72.7 in
54.4 in.
5 in. touchscreen
HEPA
184.7 cm
1
6 Years
25.7 in.
27 in.
57.2 cm
32 in.
105 W
ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
123.2 cm
Cell Logic
2 Duplex
2 Double, Left and Right Side
20 cm
1390 m³/hr.
99.99%
138.2 cm
8 in.
818 CFM
48.5 in.
22.5 in.
65.3 cm
69 cm
81.3 cm
Videos
SDS
Documents

Documents