Please login to your online account to display your discounted pricing

4-Formylphenylboronic acid, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC359380250

 View more versions of this product

Catalog No. AC359380250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (HPLC)
Formula Weight 149.94
Physical Form Liquid
Quantity 25g
Color Clear colorless to yellow-orange
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C7H7BO3
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Name Note 97%
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Chemical Name or Material 4-Formylphenylboronic acid
MDL Number MFCD00151823
CAS 87199-17-5
Recommended Storage Refrigerator +4°C

Specifications

Chemical Name or Material 4-Formylphenylboronic acid
Name Note 97%
CAS 87199-17-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C7H7BO3
Formula Weight 149.94
MDL Number MFCD00151823
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.

Refrigerator +4°C

SDS & Safety Documents