missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Formylphenyl beta-D-allopyranoside, 98%

Manufacturer:  Alfa Aesar™ J6610203

 View more versions of this product

Catalog No. AAJ6610203



Description

Description

Solubility
Soluble in hot water. Slightly soluble in water, ethanol or carbinol. Insoluble in aether or chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

4-Formylphenyl beta-D-allopyranoside
White
1g
C13H16O7
4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside, 4-formylphenyl beta-d-allopyranoside, 4-formylphenyl ss-d-allopyranoside, helicid, helicide, hilicidum
OLZAGZCCJJBKNZ-SYLRKERUSA-N
4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
12896796
Powder
−105° (c=0.8 in Water)
190°C
80154-34-3
MFCD00210992
Soluble in hot water. Slightly soluble in water,ethanol or carbinol. Insoluble in aether or chloroform.
C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
284.264
284.26
98%

Chemical Identifiers

80154-34-3
284.264
OLZAGZCCJJBKNZ-SYLRKERUSA-N
12896796
C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
C13H16O7
MFCD00210992
4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside, 4-formylphenyl beta-d-allopyranoside, 4-formylphenyl ss-d-allopyranoside, helicid, helicide, hilicidum
4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents