Please login to your online account to display your discounted pricing

4-Formylphenoxyacetic Acid 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  410780050

 View more versions of this product

Catalog No. AC410780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 180.16
Quantity 5g
Color Beige
EINECS Number 244-749-3
TSCA TSCA
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H8O4
MDL Number MFCD00016613
CAS 22042-71-3
Chemical Name or Material 4-Formylphenoxyacetic acid

Specifications

Chemical Name or Material 4-Formylphenoxyacetic acid
Name Note 99%
CAS 22042-71-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8O4
Formula Weight 180.16
MDL Number MFCD00016613