Please login to your online account to display your discounted pricing

4-Formylphenoxyacetic Acid 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Formylphenoxyacetic acid
Name Note 99%
CAS 22042-71-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8O4
Formula Weight 180.16
MDL Number MFCD00016613
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC410780250

  SDS
thermo fisher scientific
410780250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC410780050

  SDS
thermo fisher scientific
410780050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 180.16
Quantity 25g
Color Beige
EINECS Number 244-749-3
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 4-Formylphenoxyacetic acid
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H8O4
MDL Number MFCD00016613
CAS Min % 98.5
Infrared Spectrum Authentic
CAS 22042-71-3