Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Formyl-2-methoxyphenyl cyclopropanecarboxylate

$435.08 - $435.08

Chemical Identifiers

Chemical Name or Material 4-Formyl-2-methoxyphenyl cyclopropanecarboxylate
CAS 380336-99-2
Molecular Formula C12H12O4
Formula Weight 220.23g/mol
MDL Number MFCD01935794
Synonym Cyclopropanecarboxylic acid 4-formyl-2-methyoxyphenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5048606

 
alfa aesar™
H50486-06
Each for $435.08
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 4-Formyl-2-methoxyphenyl cyclopropanecarboxylate
CAS 380336-99-2
Molecular Formula C12H12O4
Formula Weight 220.23g/mol
MDL Number MFCD01935794
Synonym Cyclopropanecarboxylic acid 4-formyl-2-methyoxyphenyl ester