missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6626603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid | |
White | |
MFCD01862347,MFCD00273480 | |
7831105 | |
MLMIBGARTUSGND-UHFFFAOYSA-N | |
4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid | |
2756125 | |
98% |
188715-40-4 | |
C23H25NO4 | |
1 g | |
fmoc-4-amc-oh, fmoc-tranexamic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, n-fmoc-tranexamic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid | |
OC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1 | |
379.46 | |
379.18 | |
Powder |
Chemical Identifiers
188715-40-4 | |
379.46 | |
MLMIBGARTUSGND-UHFFFAOYSA-N | |
2756125 | |
OC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1 |
C23H25NO4 | |
MFCD01862347,MFCD00273480 | |
fmoc-4-amc-oh, fmoc-tranexamic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, n-fmoc-tranexamic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid | |
4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only