missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ H66266MD

 View more versions of this product

Catalog No. AAH66266MD



Specifications

Specifications

4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid
250mg
C23H25NO4
7831105
MLMIBGARTUSGND-UHFFFAOYSA-N
4-[(9H-fluoren-9-ylmethoxycarbonylamino)methyl]cyclohexane-1-carboxylic acid
2756125
Powder
White
188715-40-4
MFCD01862347
1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, fmoc-4-amc-oh, fmoc-tranexamic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, n-fmoc-tranexamic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid
C1CC(CCC1CNC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
379.456
379.18
98%

Chemical Identifiers

188715-40-4
379.456
MLMIBGARTUSGND-UHFFFAOYSA-N
2756125
C1CC(CCC1CNC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
C23H25NO4
MFCD01862347
1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, fmoc-4-amc-oh, fmoc-tranexamic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, n-fmoc-tranexamic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid
4-[(9H-fluoren-9-ylmethoxycarbonylamino)methyl]cyclohexane-1-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents