missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Fmoc-amino)butyric acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6614706
Description
4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
4-(Fmoc-amino)butyric acid | |
White | |
FmocNHCH2CH2CH2CO2H | |
5 g | |
fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid | |
ACUIFAAXWDLLTR-UHFFFAOYSA-N | |
4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid | |
2756086 | |
95% |
116821-47-7 | |
C19H19NO4 | |
MFCD00144889 | |
7491485 | |
Slightly soluble in water. | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O | |
325.364 | |
325.36 | |
Powder |
Chemical Identifiers
116821-47-7 | |
325.364 | |
ACUIFAAXWDLLTR-UHFFFAOYSA-N | |
2756086 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O |
C19H19NO4 | |
MFCD00144889 | |
fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid | |
4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only