missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorotoluene, TCI America™

Manufacturer:  TCI America F004125ML

 View more versions of this product

Catalog No. F004125ML



Specifications

Specifications

4-Fluorotoluene
Yellow
25mL
C7H7F
2388
WRWPPGUCZBJXKX-UHFFFAOYSA-N
1-fluoro-4-methylbenzene
9603
Clear Liquid at 20°C
116°C
-56°C
352-32-9
MFCD00000358
4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
CC1=CC=C(C=C1)F
110.131
110.13
≥99.0% (GC)

Chemical Identifiers

352-32-9
110.131
WRWPPGUCZBJXKX-UHFFFAOYSA-N
9603
CC1=CC=C(C=C1)F
C7H7F
MFCD00000358
4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
1-fluoro-4-methylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0083

RTECSNumber : XT2580000

TSCA : Yes

Documents