missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorotoluene, 99%

$48.89 - $174.26

Chemical Identifiers

CAS 352-32-9
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.131
MDL Number MFCD00000358
InChI Key WRWPPGUCZBJXKX-UHFFFAOYSA-N
Synonym 4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
PubChem CID 9603
IUPAC Name 1-fluoro-4-methylbenzene
SMILES CC1=CC=C(C=C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1036522 View Documents Alfa Aesar™
A1036522
100g Each for $48.89
 
AAA1036536 View Documents Alfa Aesar™
A1036536
500g Each for $174.26
 
Specifications

Chemical Identifiers

352-32-9
110.131
WRWPPGUCZBJXKX-UHFFFAOYSA-N
9603
CC1=CC=C(C=C1)F
C7H7F
MFCD00000358
4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
1-fluoro-4-methylbenzene

Specifications

10°C (50°F)
115°C to 116°C
100g
C7H7F
UN2388
14,4180
WRWPPGUCZBJXKX-UHFFFAOYSA-N
1-fluoro-4-methylbenzene
9603
99%
Aromatic
-56°C
352-32-9
MFCD00000358
1362373
4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
CC1=CC=C(C=C1)F
110.131
110.13
4-Fluorotoluene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H335
Highly flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
P210-P261
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES

RTECSNumber : XT2580000

TSCA : TSCA