Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorothioanisole, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluorothioanisole
CAS 371-15-3
Molecular Formula C7H7FS
Formula Weight 142.2
MDL Number MFCD00040829
UN Number UN3334
Beilstein 2041508
Synonym 4-Fluorophenyl methyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0497903

 
alfa aesar™
L0497903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.2
Chemical Name or Material 4-Fluorothioanisole
Assay 97%
MDL Number MFCD00040829
CAS 371-15-3
UN Number UN3334
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H7FS
Beilstein 2041508
Synonym 4-Fluorophenyl methyl sulfide
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Boiling Point 89° to 90°C (31mmHg)
Quantity 1g
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
Flash Point 73°C (163°F)
Odor Stench
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.