missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorosulfonyl-1-hydroxy-2-naphthoic acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics 162470100

 View more versions of this product

Catalog No. AC162470100



Specifications

Specifications

4-Fluorosulfonyl-1-hydroxy-2-naphthoic acid
Glass bottle
10g
839-78-1
100.0
C11H7FO5S
MFCD00003961
CDJFAPNPVPDTDW-UHFFFAOYSA-N
4-fluorosulfonyl-1-hydroxynaphthalene-2-carboxylic acid
70046
Amorphous Powder
Authentic
Beige to Brown
99%
98.5
99%
FSO2C10H5(OH)CO2H
4-fluorosulfonyl-1-hydroxy-2-naphthoic acid, 4-fluorosulfonyl-1-hydroxy-2-naphthoic acid, 4-fluorosulfonyl-1-hydroxynaphthalene-2-carboxylic acid, 4-fluorosulphonyl-1-hydroxy-2-naphthoic acid, 2-naphthoic acid, 4-fluorosulfonyl-1-hydroxy, 4-fluorosulfonyl-1-hydroxy-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylicacid, 4-fluorosulfonyl-1-hydroxy
C1=CC=C2C(=C1)C(=CC(=C2O)C(=O)O)S(=O)(=O)F
270.23
270.22
≥98.5%

Chemical Identifiers

839-78-1
270.23
CDJFAPNPVPDTDW-UHFFFAOYSA-N
70046
C1=CC=C2C(=C1)C(=CC(=C2O)C(=O)O)S(=O)(=O)F
C11H7FO5S
MFCD00003961
4-fluorosulfonyl-1-hydroxy-2-naphthoic acid, 4-fluorosulfonyl-1-hydroxy-2-naphthoic acid, 4-fluorosulfonyl-1-hydroxynaphthalene-2-carboxylic acid, 4-fluorosulphonyl-1-hydroxy-2-naphthoic acid, 2-naphthoic acid, 4-fluorosulfonyl-1-hydroxy, 4-fluorosulfonyl-1-hydroxy-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylicacid, 4-fluorosulfonyl-1-hydroxy
4-fluorosulfonyl-1-hydroxynaphthalene-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 212-659-3

SDS Documents