Please login to your online account to display your discounted pricing

4-Fluorosulfonyl-1-hydroxy-2-naphthoic acid, 99%, ACROS Organics™

$217.43 - $217.43

Chemical Identifiers

Chemical Name or Material 4-Fluorosulfonyl-1-hydroxy-2-naphthoic acid
Name Note 99%
CAS 839-78-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11 H7 F O5 S
Linear Formula FSO2C10H5(OH)CO2H
Formula Weight 270.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC162470100

  SDS
thermo fisher scientific
162470100
Glass bottle 10g Each for $217.43
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Amorphous Powder
Formula Weight 270.22
Quantity 10g
Color Beige to brown
EINECS Number 212-659-3
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Assay Percent Range 99%
Chemical Name or Material 4-Fluorosulfonyl-1-hydroxy-2-naphthoic acid
MDL Number MFCD00003961
Linear Formula FSO2C10H5(OH)CO2H
Molecular Formula C11 H7 F O5 S
Name Note 99%
CAS 839-78-1
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.