Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-4-Fluorostyrene oxide, 98%

Chemical Identifiers

Chemical Name or Material (+/-)-4-Fluorostyrene oxide
CAS 18511-62-1
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD00043994
UN Number UN2810
Synonym (+/-)-2-(4-Fluorophenyl)oxirane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2660006

  SDS
alfa aesar™
H2660006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2660014

  SDS
alfa aesar™
H2660014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.14
Chemical Name or Material (+/-)-4-Fluorostyrene oxide
Assay 98%
Molecular Formula C8H7FO
MDL Number MFCD00043994
CAS 18511-62-1
UN Number UN2810
Synonym (+/-)-2-(4-Fluorophenyl)oxirane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H312-H332-H319-H227
Suspected of causing cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Flash Point 77°C (171°F)
Odor Pungent
Boiling Point 92°C (14mmHg)
Quantity 5g

Applications
(+/-)-4-Fluorostyrene oxide used in chemicals, pharmaceutical intermediates. It is also used in organic chemistry that has also many industrial applications, biotransformation

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H312-H332-H319-H227
Suspected of causing cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Combustible liquid.