Please login to your online account to display your discounted pricing

4-Fluorostyrene, 97%, stabilized, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC168340500

 View more versions of this product

Catalog No. AC168340500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 122.14
Quantity 50g
Color Clear colorless to very slightly yellow
Density 1.024g/mL
CAS Min % 96.0
CAS Max % 100.0
Linear Formula H2C=CHC6H4F
Infrared Spectrum Authentic
Name Note Stabilized
Assay Percent Range 97%
Chemical Name or Material 4-Fluorostyrene
CAS 405-99-2
Molecular Formula C8H7F
Health Hazard 1 GHS Signal Word: Warning
CAS 98-29-3
MDL Number MFCD00000361
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
Packaging Glass bottle
Stabilizer 0.1% tert.-butylcatechol
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Fluorostyrene
Name Note Stabilized
CAS 405-99-2
CAS Min % 96.0
CAS Max % 100.0
CAS 98-29-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7F
Linear Formula H2C=CHC6H4F
Formula Weight 122.14
MDL Number MFCD00000361
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents