missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorosalicylaldehyde, 98%

$296.19 - $296.19

Chemical Identifiers

CAS 348-28-7
Molecular Formula C7H5FO2
Formula Weight 140.12
MDL Number MFCD03788526
Synonym 4-Fluoro-2-hydroxybenzaldehyde
Chemical Name or Material 4-Fluorosalicylaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2614903 SDS Alfa Aesar™
H2614903
1g Each for $296.19
Specifications Description & Specifications

Chemical Identifiers

348-28-7
140.12
4-Fluoro-2-hydroxybenzaldehyde
C7H5FO2
MFCD03788526
4-Fluorosalicylaldehyde

Specifications

70° to 72°C
348-28-7
140.12
4-Fluoro-2-hydroxybenzaldehyde
98%
1g
C7H5FO2
MFCD03788526
4-Fluorosalicylaldehyde

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning