missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenylurea, 96%

$58.92 - $205.11

Chemical Identifiers

CAS 659-30-3
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.144
MDL Number MFCD00014787
InChI Key IQZBVVPYTDHTIP-UHFFFAOYSA-N
Synonym 4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, n-4-fluorophenyl urea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea
PubChem CID 12612
IUPAC Name (4-fluorophenyl)urea
SMILES C1=CC(=CC=C1NC(=O)N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0131406 View Documents Alfa Aesar™
L0131406
5g Each for $58.92
 
AAL0131414 View Documents Alfa Aesar™
L0131414
25g Each for $205.11
 
Description

Description

Applications
4-Fluorophenylurea is widely used as a filter for drug.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

659-30-3
154.144
IQZBVVPYTDHTIP-UHFFFAOYSA-N
12612
C1=CC(=CC=C1NC(=O)N)F
C7H7FN2O
MFCD00014787
4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, n-4-fluorophenyl urea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea
(4-fluorophenyl)urea

Specifications

183°C to 185°C
659-30-3
MFCD00014787
4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, n-4-fluorophenyl urea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea
C1=CC(=CC=C1NC(=O)N)F
154.144
154.14
4-Fluorophenylurea
5g
C7H7FN2O
2090131
IQZBVVPYTDHTIP-UHFFFAOYSA-N
(4-fluorophenyl)urea
12612
96%
Safety and Handling

Safety and Handling

RTECSNumber : YT3980000