Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Fluorophenylthio)acetonitrile, 97%

$230.68 - $230.68

Chemical Identifiers

Chemical Name or Material (4-Fluorophenylthio)acetonitrile
CAS 18527-21-4
Molecular Formula C8H6FNS
Formula Weight 167.21g/mol
MDL Number MFCD00179342
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2052603

 
alfa aesar™
B20526-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2052606

 
alfa aesar™
B20526-06
5g Each for $230.68
Description & Specifications

Specifications

Melting Point 32° to 35°C
Quantity 1g
Odor Unpleasant
Chemical Name or Material (4-Fluorophenylthio)acetonitrile
CAS 18527-21-4
Assay 97%
Molecular Formula C8H6FNS
Formula Weight 167.21g/mol
MDL Number MFCD00179342
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.