Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Fluorophenylthio)acetone, 97%

$45.09 - $45.09

Chemical Identifiers

Chemical Name or Material (4-Fluorophenylthio)acetone
CAS 2968-13-0
Molecular Formula C9H9FOS
Formula Weight 184.23
MDL Number MFCD00179340
Synonym (4-Fluorophenylthio)propan-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2009403

 
alfa aesar™
B2009403
1g Each for $45.09
Description & Specifications

Specifications

Formula Weight 184.23
Chemical Name or Material (4-Fluorophenylthio)acetone
Assay 97%
Molecular Formula C9H9FOS
MDL Number MFCD00179340
CAS 2968-13-0
Synonym (4-Fluorophenylthio)propan-2-one
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 100° to 103°C (1mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.