Please login to your online account to display your discounted pricing

4-Fluorophenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC377371000

 View more versions of this product

Catalog No. AC377371000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 199.3
Quantity 100mL
Color Clear yellow to red-brown
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 2 to 2.2M
Density 1.0430g/mL
Chemical Name or Material 4-Fluorophenylmagnesium bromide
Molecular Formula C6H4BrFMg
CAS 352-13-6
Health Hazard 2 GHS P Statement Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gas. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. Reacts violently with water.
CAS 60-29-7
CAS Min % 57.96
Name Note 2M Solution in Diethyl Ether
Linear Formula F(C6H4)MgBr
MDL Number MFCD00009667
CAS Min % 38.22
Health Hazard 1 Danger
Additional Information product may develop some turbidity or precipitate
CAS Max % 42.04
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
CAS Max % 61.78

Specifications

Chemical Name or Material 4-Fluorophenylmagnesium bromide
Name Note 2M Solution in Diethyl Ether
CAS 352-13-6
CAS Min % 38.22
CAS Max % 42.04
CAS 60-29-7
CAS Min % 57.96
CAS Max % 61.78
Molecular Formula C6H4BrFMg
Linear Formula F(C6H4)MgBr
Formula Weight 199.3
MDL Number MFCD00009667
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS P Statement Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gas. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. Reacts violently with water.

SDS & Safety Documents