Learn More
4-Fluorophenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 377378000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Fluorophenylmagnesium bromide | |
352-13-6,60-29-7 | |
42.04,61.78 | |
F(C6H4)MgBr | |
2 to 2.2M | |
4-fluorophenylmagnesium bromide, grignard reagent, bromo 4-fluorophenyl magnesium, bromo 4-fluorophenyl magnesium, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorophenylmagnesiumbromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, 4-fluorophenyl magnesiumbromide | |
C1=CC(=CC=[C-]1)F.[Mg+2].[Br-] | |
199.305 | |
199.3 | |
800mL |
2M Solution in Diethyl Ether | |
38.22,57.96 | |
C6H4BrFMg | |
MFCD00009667 | |
1.0430g/mL | |
BRKADVNLTRCLOW-UHFFFAOYSA-M | |
magnesium;fluorobenzene;bromide | |
2734897 | |
AcroSeal™ Glass Bottle |
Chemical Identifiers
352-13-6 | |
199.305 | |
BRKADVNLTRCLOW-UHFFFAOYSA-M | |
2734897 | |
C1=CC(=CC=[C-]1)F.[Mg+2].[Br-] |
C6H4BrFMg | |
MFCD00009667 | |
4-fluorophenylmagnesium bromide, grignard reagent, bromo 4-fluorophenyl magnesium, bromo 4-fluorophenyl magnesium, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorophenylmagnesiumbromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, 4-fluorophenyl magnesiumbromide | |
magnesium;fluorobenzene;bromide |
Safety and Handling
GHS P Statement Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gas. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. Reacts violently with water.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Danger
RUO â Research Use Only