Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenylmagnesium bromide, 1M in MeTHF

Chemical Identifiers

Chemical Name or Material 4-Fluorophenylmagnesium bromide
CAS 352-13-6
Molecular Formula C6H4BrFMg
Formula Weight 199.31
MDL Number MFCD00009667
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51169AE

 
alfa aesar™
H51169AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH51169AP

 
alfa aesar™
H51169AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.31
Chemical Name or Material 4-Fluorophenylmagnesium bromide
TSCA TSCA
Molecular Formula C6H4BrFMg
MDL Number MFCD00009667
CAS 352-13-6
UN Number UN2924
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Concentration 1M in MeTHF
Quantity 100mL

Applications
4-Fluorophenylmagnesium bromide is used as a reagent for the introduction of the 4-Fluorophenyl group.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.