Please login to your online account to display your discounted pricing

4-Fluorophenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC377238000

 View more versions of this product

Catalog No. AC377238000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 199.3
Quantity 800mL
Color Clear yellow to brown
Packaging AcroSeal™ Glass Bottle
Assay 14 to 17%
Density 1.0100g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. In contact with water releases flammable gas. Reacts violently with water. May form explosive peroxides.
Chemical Name or Material 4-Fluorophenylmagnesium bromide
Name Note 0.8M Solution in THF
Molecular Formula C6H4BrFMg
Linear Formula F(C6H6)MgBr
MDL Number MFCD00009667
CAS 352-13-6
CAS 109-99-9
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Storage Information product may develop some turbidity or precipitate during storage

Specifications

Chemical Name or Material 4-Fluorophenylmagnesium bromide
Name Note 0.8M Solution in THF
CAS 352-13-6
CAS 109-99-9
Assay 14 to 17%
Molecular Formula C6H4BrFMg
Linear Formula F(C6H6)MgBr
Formula Weight 199.3
MDL Number MFCD00009667
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. In contact with water releases flammable gas. Reacts violently with water. May form explosive peroxides.

SDS & Safety Documents