missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorophenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

CAS 352-13-6,109-99-9
Molecular Formula C6H4BrFMg
Linear Formula F(C6H6)MgBr
Formula Weight 199.3
MDL Number MFCD00009667
Chemical Name or Material 4-Fluorophenylmagnesium bromide, 0.8M Solution in THF
Assay 14 to 17%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC377238000 SDS Thermo Fisher Scientific
AC377238000
AcroSeal™ Glass Bottle 800mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC377231000 SDS Thermo Fisher Scientific
AC377231000
AcroSeal™ Glass Bottle in Tin Can 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

352-13-6,109-99-9
F(C6H6)MgBr
MFCD00009667
14 to 17%
C6H4BrFMg
199.3
4-Fluorophenylmagnesium bromide, 0.8M Solution in THF

Specifications

199.3
800mL
AcroSeal™ Glass Bottle
109-99-9,352-13-6
F(C6H6)MgBr
C6H4BrFMg
14 to 17%
Solution
Clear yellow to brown
1.0100g/mL
product may develop some turbidity or precipitate during storage
MFCD00009667
4-Fluorophenylmagnesium bromide, 0.8M Solution in THF

Safety and Handling

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. In contact with water releases flammable gas. Reacts violently with water. May form explosive peroxides.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger