missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorophenylacetyl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC376120050

 View more versions of this product

Catalog No. AC376120050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Fluorophenylacetyl chloride
107°C
Glass bottle
1.259
Yellow
459-04-1
100.0
C8H6ClFO
MFCD00674181
Solubility in water: reacts
C1=CC(=CC=C1CC(=O)Cl)F
172.59
172.59
98%
1.2590g/mL
Authentic
1.5110 to 1.513
60°C (8.0mmHg)
5g
97.5
97.5% min. (GC)
FC6H4CH2C(O)Cl
2-4-fluorophenyl acetyl chloride, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, 4-fluorobenzeneacetyl chloride, 4-fluorophenylacetyl chloride, 4-fluorophenylacetylchloride, acmc-1aivq, benzeneacetyl chloride, 4-fluoro, p-fluorophenylacetyl chloride, pubchem17517
SIOJFYRPBYGHOO-UHFFFAOYSA-N
2-(4-fluorophenyl)acetyl chloride
3744988
Liquid

Chemical Identifiers

459-04-1
172.59
SIOJFYRPBYGHOO-UHFFFAOYSA-N
3744988
C1=CC(=CC=C1CC(=O)Cl)F
C8H6ClFO
MFCD00674181
2-4-fluorophenyl acetyl chloride, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, 4-fluorobenzeneacetyl chloride, 4-fluorophenylacetyl chloride, 4-fluorophenylacetylchloride, acmc-1aivq, benzeneacetyl chloride, 4-fluoro, p-fluorophenylacetyl chloride, pubchem17517
2-(4-fluorophenyl)acetyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents