Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenylacetyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluorophenylacetyl chloride
CAS 459-04-1
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD00674181
UN Number UN3265
Beilstein 972148
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1923503

 
alfa aesar™
L1923503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1923514

 
alfa aesar™
L1923514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1923506

 
alfa aesar™
L1923506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.59
MDL Number MFCD00674181
CAS 459-04-1
Beilstein 972148
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 4-Fluorophenylacetyl chloride
Molecular Formula C8H6ClFO
UN Number UN3265
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 107°C (224°F)
Quantity 1g
Assay 97%
Boiling Point 60°C (8mmHg)

Applications
4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

Solubility
Miscible with dichloromethane.

Notes
Moisture Sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.