missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorophenylacetone, 99%, Thermo Scientific Chemicals
$56.42 - $184.03
Chemical Identifiers
CAS | 459-03-0 |
---|---|
Molecular Formula | C9H9FO |
Molecular Weight (g/mol) | 152.17 |
MDL Number | MFCD00000362 |
InChI Key | ZUEKIIWSVFBTCM-UHFFFAOYSA-N |
Synonym | 4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one |
PubChem CID | 521187 |
IUPAC Name | 1-(4-fluorophenyl)propan-2-one |
SMILES | CC(=O)CC1=CC=C(F)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1172003
|
Thermo Scientific Chemicals
L1172003 |
1 g |
Each for $56.42
|
|
|||||
AAL1172006
|
Thermo Scientific Chemicals
L1172006 |
5 g |
Each for $184.03
|
|
|||||
Description
4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
459-03-0 | |
152.17 | |
ZUEKIIWSVFBTCM-UHFFFAOYSA-N | |
521187 | |
CC(=O)CC1=CC=C(F)C=C1 |
C9H9FO | |
MFCD00000362 | |
4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one | |
1-(4-fluorophenyl)propan-2-one |
Specifications
459-03-0 | |
106°C to 107°C (18 mmHg) | |
C9H9FO | |
MFCD00000362 | |
1936697 | |
Not miscible or difficult to mix with water. | |
CC(=O)CC1=CC=C(F)C=C1 | |
152.17 | |
152.17 | |
4-Fluorophenylacetone |
1.11 | |
91°C (195°F) | |
1.4958 | |
1 g | |
4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one | |
ZUEKIIWSVFBTCM-UHFFFAOYSA-N | |
1-(4-fluorophenyl)propan-2-one | |
521187 | |
99% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 207-284-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only