missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorophenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America F020625G
Specifications
4-Fluorophenylacetic Acid | |
85°C | |
164°C | |
MFCD00004343 | |
4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid | |
C1=CC(=CC=C1CC(=O)O)F | |
154.14 | |
154.14 | |
Crystalline Powder |
405-50-5 | |
White | |
C8H7FO2 | |
25 g | |
MGKPFALCNDRSQD-UHFFFAOYSA-N | |
2-(4-fluorophenyl)acetic acid | |
9837 | |
≥98.0% (T) |
Chemical Identifiers
405-50-5 | |
154.14 | |
MGKPFALCNDRSQD-UHFFFAOYSA-N | |
9837 | |
C1=CC(=CC=C1CC(=O)O)F |
C8H7FO2 | |
MFCD00004343 | |
4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid | |
2-(4-fluorophenyl)acetic acid |
Safety and Handling
TSCA : Yes