Alfa Aesar™ 4-Fluorophenyl trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate, 97%
Manufacturer:  Alfa Aesar™ H5573303
 View more versions of this product
Catalog No. AAH5573303
Specifications
Specifications
4-Fluorophenyl trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate
1g
C24H35FO2
1,1'-bicyclohexyl-4-carboxylic acid, 4'-pentyl-, 4-fluorophenyl ester
CCCCCC1CCC(CC1)C2CCC(CC2)C(=O)OC3=CC=C(C=C3)F
374.54
374.54
64°C to 66°C
88878-50-6
MFCD11053423
ZVCPJOLFAYLVER-UHFFFAOYSA-N
(4-fluorophenyl) 4-(4-pentylcyclohexyl)cyclohexane-1-carboxylate
557929
97%
Chemical Identifiers
88878-50-6
374.54
ZVCPJOLFAYLVER-UHFFFAOYSA-N
557929
CCCCCC1CCC(CC1)C2CCC(CC2)C(=O)OC3=CC=C(C=C3)F
C24H35FO2
MFCD11053423
1,1'-bicyclohexyl-4-carboxylic acid, 4'-pentyl-, 4-fluorophenyl ester
(4-fluorophenyl) 4-(4-pentylcyclohexyl)cyclohexane-1-carboxylate
Documents