Please login to your online account to display your discounted pricing

4-Fluorophenyl methyl sulfone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluorophenyl methyl sulfone
Name Note 99%
CAS 455-15-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7FO2S
Linear Formula FC6H4SOCH3
Formula Weight 174.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303360010

  SDS
thermo fisher scientific
AC303360010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Adhering Crystalline Powder
Formula Weight 174.19
Quantity 1g
Color Almost white
EINECS Number 207-237-0
Melting Point 77° to 80°C
CAS Max % 100.0
MDL Number MFCD00039753
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Fluorophenyl methyl sulfone
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H7FO2S
Linear Formula FC6H4SOCH3
CAS 455-15-2
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.