Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenyl methyl sulfone, 98%

$122.38 - $122.38

Chemical Identifiers

Chemical Name or Material 4-Fluorophenyl methyl sulfone
CAS 455-15-2
Molecular Formula C7H7FO2S
Formula Weight 174.19
MDL Number MFCD00039753
Beilstein 2045674
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1286306

 
alfa aesar™
A1286306
5g Each for $122.38
Description & Specifications

Specifications

Formula Weight 174.19
Chemical Name or Material 4-Fluorophenyl methyl sulfone
Assay 98%
Molecular Formula C7H7FO2S
MDL Number MFCD00039753
CAS 455-15-2
Beilstein 2045674
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 76° to 81°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.