missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorophenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ A1556114

 View more versions of this product

Catalog No. AAA1556114



Specifications

Specifications

4-Fluorophenyl isothiocyanate
228°C
25g
C7H4FNS
UN2923
4-fluorophenyl isothiocyanate, p-fluorophenyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanato, 4-fluorophenylisothiocyanate, 1-fluoro-4-isothiocyanato-benzene, isothiocyanic acid, p-fluorophenyl ester, isothiocyanic acid 4-fluorophenyl ester, 4-fluorobenzenisothiocyanate, 4-fluoroisothiocyanatobenzene, pubchem5008
C1=CC(=CC=C1N=C=S)F
153.174
153.18
89°C (192°F)
24°C to 28°C
1544-68-9
MFCD00004809
636596
NFIUJHJMCQQYDL-UHFFFAOYSA-N
1-fluoro-4-isothiocyanatobenzene
15241
98%

Chemical Identifiers

1544-68-9
153.174
NFIUJHJMCQQYDL-UHFFFAOYSA-N
15241
C1=CC(=CC=C1N=C=S)F
C7H4FNS
MFCD00004809
4-fluorophenyl isothiocyanate, p-fluorophenyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanato, 4-fluorophenylisothiocyanate, 1-fluoro-4-isothiocyanato-benzene, isothiocyanic acid, p-fluorophenyl ester, isothiocyanic acid 4-fluorophenyl ester, 4-fluorobenzenisothiocyanate, 4-fluoroisothiocyanatobenzene, pubchem5008
1-fluoro-4-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NX8760000

TSCA : TSCA

Documents